Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736585.2 | XP_014592071.2 | 3355 | 65 | ||
XM_023630593.1 | XP_023486361.1 | 3355 | 65 | |||
XM_023630594.1 | XP_023486362.1 | 3606 | 65 | |||
XM_023630595.1 | XP_023486363.1 | 4575 | 65 | |||
XM_023630596.1 | XP_023486364.1 | 3355 | 65 | |||
XM_023630597.1 | XP_023486365.1 | 3161 | 65 | |||
XM_023630598.1 | XP_023486366.1 | 3442 | 65 | |||
XM_023630599.1 | XP_023486367.1 | 3133 | 65 | |||
XM_023630600.1 | XP_023486368.1 | 2905 | 65 | |||
XM_023630601.1 | XP_023486369.1 | 5746 | 65 | |||
XM_023630602.1 | XP_023486370.1 | 2394 | 65 | |||
XM_023630603.1 | XP_023486371.1 | 2334 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
