Assay Details
Gene Symbol: ERI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915823.4 | XP_001915858.1 | 854 | 93 | ||
XM_023630815.1 | XP_023486583.1 | 832 | 93 | |||
XM_023630816.1 | XP_023486584.1 | 856 | 93 | |||
XM_023630817.1 | XP_023486585.1 | 871 | 93 | |||
XM_023630818.1 | XP_023486586.1 | 607 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
