Assay Details
Gene Symbol: VEGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630689.1 | XP_023486457.1 | 570 | 77 | ||
XM_023630690.1 | XP_023486458.1 | 2640 | 77 | |||
XM_023630691.1 | XP_023486459.1 | 602 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
