Assay Details
Gene Symbol: NUDT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495348.6 | XP_001495398.5 | 705 | 185 | ||
XM_005606503.3 | XP_005606560.2 | 708 | 185 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
