Assay Details
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631867.1 | XP_023487635.1 | 79 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023631867.1 transcripts.
Set Membership: |
![]() ![]() |
