Assay Details
Gene Symbol: TCP11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631489.1 | XP_023487257.1 | 1102 | 73 | ||
XM_023631490.1 | XP_023487258.1 | 1241 | 73 | |||
XM_023631491.1 | XP_023487259.1 | 1125 | 73 | |||
XM_023631492.1 | XP_023487260.1 | 987 | 73 | |||
XM_023631493.1 | XP_023487261.1 | 917 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
