Assay Details
Gene Symbol: RAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500793.5 | XP_001500843.1 | 358 | 81 | ||
XM_023631107.1 | XP_023486875.1 | 517 | 81 | |||
XM_023631108.1 | XP_023486876.1 | 361 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
