Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631103.1 | XP_023486871.1 | 946 | 107 | ||
XM_023631104.1 | XP_023486872.1 | 3136 | 107 | |||
XM_023631105.1 | XP_023486873.1 | 838 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
