Assay Details
Gene Symbol: JOSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501639.4 | XP_001501689.1 | 1324 | 72 | ||
XM_023631656.1 | XP_023487424.1 | 1229 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
