Assay Details
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606645.3 | XP_005606702.1 | 101 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005606645.3 transcripts.
Set Membership: |
![]() ![]() |
