Assay Details
Gene Symbol: PMM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502546.6 | XP_001502596.1 | 447 | 72 | ||
XM_023631718.1 | XP_023487486.1 | 471 | 72 | |||
XM_023631719.1 | XP_023487487.1 | 471 | 72 | |||
XM_023631720.1 | XP_023487488.1 | 447 | 72 | |||
XM_023631721.1 | XP_023487489.1 | 1405 | 72 | |||
XM_023631722.1 | XP_023487490.1 | 468 | 72 | |||
XM_023631723.1 | XP_023487491.1 | 1335 | 72 | |||
XM_023631724.1 | XP_023487492.1 | 1715 | 72 | |||
XM_023631726.1 | XP_023487494.1 | 492 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
