Assay Details
Gene Symbol: SEPTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606696.3 | XP_005606753.3 | 1853 | 62 | ||
XM_014736869.2 | XP_014592355.2 | 1853 | 62 | |||
XM_023631047.1 | XP_023486815.1 | 1854 | 62 | |||
XM_023631048.1 | XP_023486816.1 | 1853 | 62 | |||
XM_023631049.1 | XP_023486817.1 | 504 | 62 | |||
XM_023631050.1 | XP_023486818.1 | 586 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
