Assay Details
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500690.5 | XP_001500740.3 | 1023 | 73 | ||
XM_014736875.2 | XP_014592361.1 | 1023 | 73 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500690.5 | XP_001500740.3 | 1023 | 73 | ||
XM_014736875.2 | XP_014592361.1 | 1023 | 73 |