Assay Details
Gene Symbol: TTLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631851.1 | XP_023487619.1 | 989 | 85 | ||
XM_023631852.1 | XP_023487620.1 | 1000 | 85 | |||
XM_023631853.1 | XP_023487621.1 | 1083 | 85 | |||
XM_023631854.1 | XP_023487622.1 | 1094 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
