Assay Details
Gene Symbol: PNPLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606765.3 | XP_005606822.3 | 743 | 67 | ||
XM_014736936.2 | XP_014592422.2 | 743 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
