Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737059.2 | XP_014592545.2 | 2427 | 59 | ||
XM_023632280.1 | XP_023488048.1 | 2575 | 59 | |||
XM_023632281.1 | XP_023488049.1 | 2309 | 59 | |||
XM_023632282.1 | XP_023488050.1 | 2447 | 59 | |||
XM_023632283.1 | XP_023488051.1 | 2542 | 59 | |||
XM_023632284.1 | XP_023488052.1 | 2527 | 59 | |||
XM_023632285.1 | XP_023488053.1 | 2497 | 59 | |||
XM_023632286.1 | XP_023488054.1 | 2494 | 59 | |||
XM_023632287.1 | XP_023488055.1 | 2479 | 59 | |||
XM_023632288.1 | XP_023488056.1 | 2475 | 59 | |||
XM_023632289.1 | XP_023488057.1 | 2209 | 59 | |||
XM_023632290.1 | XP_023488058.1 | 2449 | 59 | |||
XM_023632291.1 | XP_023488059.1 | 2129 | 59 | |||
XM_023632293.1 | XP_023488061.1 | 2144 | 59 | |||
XM_023632294.1 | XP_023488062.1 | 2025 | 59 | |||
XM_023632295.1 | XP_023488063.1 | 2005 | 59 | |||
XM_023632296.1 | XP_023488064.1 | 1972 | 59 | |||
XM_023632297.1 | XP_023488065.1 | 1957 | 59 | |||
XM_023632298.1 | XP_023488066.1 | 1924 | 59 | |||
XM_023632299.1 | XP_023488067.1 | 1878 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
