Assay Details
Gene Symbol: NUDT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499156.5 | XP_001499206.1 | 546 | 54 | ||
XM_005606924.3 | XP_005606981.1 | 550 | 54 | |||
XM_023631943.1 | XP_023487711.1 | 438 | 54 | |||
GenBank mRNA | JL638068.1 | - | 4 - 5 | 263 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
