Assay Details
Gene Symbol: ATP5F1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499861.6 | XP_001499911.3 | 736 | 72 | ||
XM_005606935.3 | XP_005606992.1 | 736 | 72 | |||
XM_005606936.3 | XP_005606993.1 | 736 | 72 | |||
GenBank mRNA | JL626894.1 | - | 5 - 6 | 589 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
