Assay Details
Gene Symbol: TASOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606953.3 | XP_005607010.1 | 8065 | 114 | ||
XM_005606954.3 | XP_005607011.1 | 7667 | 114 | |||
XM_014737100.2 | XP_014592586.2 | 8960 | 114 | |||
XM_014737101.2 | XP_014592587.2 | 8960 | 114 | |||
XM_014737102.2 | XP_014592588.2 | 8961 | 114 | |||
XM_014737104.2 | XP_014592590.1 | 7124 | 114 | |||
XM_023632334.1 | XP_023488102.1 | 7833 | 114 | |||
XM_023632335.1 | XP_023488103.1 | 8961 | 114 | |||
XM_023632336.1 | XP_023488104.1 | 8960 | 114 | |||
XM_023632338.1 | XP_023488106.1 | 8718 | 114 | |||
XM_023632339.1 | XP_023488107.1 | 7806 | 114 |

More Information
Set Membership: |
![]() ![]() |
