Assay Details
Gene Symbol: AKR1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737113.2 | XP_014592599.1 | 1231 | 71 | ||
XM_023632188.1 | XP_023487956.1 | 1408 | 71 | |||
XM_023632189.1 | XP_023487957.1 | 1405 | 71 | |||
XM_023632190.1 | XP_023487958.1 | 1354 | 71 | |||
XM_023632192.1 | XP_023487960.1 | 1324 | 71 | |||
XM_023632193.1 | XP_023487961.1 | 1236 | 71 | |||
XM_023632194.1 | XP_023487962.1 | 1295 | 71 | |||
XM_023632195.1 | XP_023487963.1 | 5746 | 71 | |||
XM_023632196.1 | XP_023487964.1 | 4649 | 71 | |||
XM_023632197.1 | XP_023487965.1 | 5022 | 71 | |||
XM_023632198.1 | XP_023487966.1 | 4975 | 71 | |||
XM_023632199.1 | XP_023487967.1 | 4768 | 71 | |||
XM_023632200.1 | XP_023487968.1 | 5828 | 71 | |||
XM_023632201.1 | XP_023487969.1 | 5836 | 71 | |||
XM_023632202.1 | XP_023487970.1 | 8767 | 71 | |||
XM_023632203.1 | XP_023487971.1 | 8482 | 71 | |||
XM_023632204.1 | XP_023487972.1 | 8573 | 71 | |||
XM_023632205.1 | XP_023487973.1 | 8519 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
