Assay Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637028.1 | XP_023492796.1 | 680 | 68 | ||
XM_023637029.1 | XP_023492797.1 | 677 | 68 | |||
XM_023637030.1 | XP_023492798.1 | 682 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
