Assay Details
Gene Symbol: ADGRG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608284.3 | XP_005608341.1 | 1104 | 58 | ||
XM_014738454.2 | XP_014593940.1 | 1216 | 58 | |||
XM_014738455.2 | XP_014593941.1 | 1106 | 58 | |||
XM_014738456.2 | XP_014593942.1 | 988 | 58 | |||
XM_014738457.2 | XP_014593943.1 | 619 | 58 | |||
XM_023637053.1 | XP_023492821.1 | 1064 | 58 | |||
XM_023637054.1 | XP_023492822.1 | 1217 | 58 | |||
XM_023637055.1 | XP_023492823.1 | 959 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
