Assay Details
Gene Symbol: ADGRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608288.2 | XP_005608345.1 | 445 | 66 | ||
XM_005608290.3 | XP_005608347.1 | 446 | 66 | |||
XM_023637062.1 | XP_023492830.1 | 693 | 66 | |||
XM_023637063.1 | XP_023492831.1 | 445 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
