Assay Details
Gene Symbol: BEAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608306.3 | XP_005608363.1 | 557 | 82 | ||
XM_014738450.2 | XP_014593936.1 | 433 | 82 | |||
XM_023637121.1 | XP_023492889.1 | 559 | 82 | |||
XM_023637123.1 | XP_023492891.1 | 547 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005608306.3, XM_014738450.2, XM_023637121.1, XM_023637123.1 transcripts.
Set Membership: |
![]() ![]() |
