Assay Details
Gene Symbol: BEAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738450.2 | XP_014593936.1 | 542 | 77 | ||
XM_023637122.1 | XP_023492890.1 | 246 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014738450.2, XM_023637122.1 transcripts.
Set Membership: |
![]() ![]() |
