Assay Details
Gene Symbol: BEAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608306.3 | XP_005608363.1 | 1196 | 55 | ||
XM_014738450.2 | XP_014593936.1 | 952 | 55 | |||
XM_023637120.1 | XP_023492888.1 | 776 | 55 | |||
XM_023637121.1 | XP_023492889.1 | 1195 | 55 | |||
XM_023637122.1 | XP_023492890.1 | 656 | 55 | |||
XM_023637123.1 | XP_023492891.1 | 1063 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
