Assay Details
Gene Symbol: CMTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495994.4 | XP_001496044.1 | 647 | 119 | ||
XM_005608311.3 | XP_005608368.1 | 488 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
