Assay Details
Gene Symbol: RIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638631.1 | XP_023494399.1 | 1832 | 59 | ||
XM_023638632.1 | XP_023494400.1 | 1805 | 59 | |||
XM_023638633.1 | XP_023494401.1 | 1798 | 59 | |||
XM_023638634.1 | XP_023494402.1 | 2043 | 59 | |||
XM_023638635.1 | XP_023494403.1 | 1754 | 59 | |||
XM_023638636.1 | XP_023494404.1 | 1568 | 59 | |||
XM_023638637.1 | XP_023494405.1 | 1733 | 59 | |||
XM_023638638.1 | XP_023494406.1 | 1832 | 59 | |||
XM_023638639.1 | XP_023494407.1 | 2343 | 59 | |||
XM_023638640.1 | XP_023494408.1 | 1105 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
