Assay Details
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498476.4 | XP_001498526.1 | 658 | 68 | ||
XM_005608347.3 | XP_005608404.1 | 908 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
