Assay Details
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608382.3 | XP_005608439.1 | 965 | 62 | ||
XM_014738441.2 | XP_014593927.1 | 965 | 62 | |||
XM_014738442.2 | XP_014593928.1 | 901 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
