Assay Details
Gene Symbol: ZNRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608432.3 | XP_005608489.2 | 1152 | 59 | ||
XM_023637332.1 | XP_023493100.1 | 1151 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
