Assay Details
Gene Symbol: CHST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608436.3 | XP_005608493.1 | 7454 | 103 | ||
XM_005608437.3 | XP_005608494.1 | 3007 | 103 | |||
XM_023637343.1 | XP_023493111.1 | 3154 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005608437.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
