Assay Details
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502034.6 | XP_001502084.1 | 343 | 97 | ||
GenBank mRNA | JL615870.1 | - | 3 - 4 | 292 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
