Assay Details
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637508.1 | XP_023493276.1 | 536 | 57 | ||
XM_023637509.1 | XP_023493277.1 | 404 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
