Assay Details
Gene Symbol: DTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611346.3 | XP_005611403.2 | 449 | 65 | ||
XM_005611348.3 | XP_005611405.2 | 441 | 65 | |||
XM_014740943.2 | XP_014596429.1 | 894 | 65 | |||
XM_014740944.2 | XP_014596430.1 | 886 | 65 | |||
XM_023643882.1 | XP_023499650.1 | 837 | 65 | |||
XM_023643883.1 | XP_023499651.1 | 641 | 65 | |||
XM_023643884.1 | XP_023499652.1 | 502 | 65 | |||
XM_023643885.1 | XP_023499653.1 | 520 | 65 | |||
XM_023643886.1 | XP_023499654.1 | 516 | 65 | |||
XM_023643887.1 | XP_023499655.1 | 494 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005611346.3, XM_005611348.3, XM_014740943.2, XM_014740944.2, XM_023643882.1, XM_023643883.1, XM_023643884.1, XM_023643885.1, XM_023643886.1, XM_023643887.1 transcripts.
Set Membership: |
![]() ![]() |
