Assay Details
Gene Symbol: DTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611348.3 | XP_005611405.2 | 530 | 66 | ||
XM_014740944.2 | XP_014596430.1 | 975 | 66 | |||
XM_023643887.1 | XP_023499655.1 | 583 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005611348.3, XM_014740944.2, XM_023643887.1 transcripts.
Set Membership: |
![]() ![]() |
