Assay Details
Gene Symbol: TSFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490176.4 | XP_001490226.1 | 578 | 74 | ||
XM_014740887.2 | XP_014596373.1 | 644 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
