Assay Details
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643900.1 | XP_023499668.1 | 333 | 88 | ||
XM_023643901.1 | XP_023499669.1 | 333 | 88 | |||
XM_023643902.1 | XP_023499670.1 | 172 | 88 | |||
XM_023643903.1 | XP_023499671.1 | 44 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023643903.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
