Assay Details
Gene Symbol: TMBIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491112.4 | XP_001491162.1 | 847 | 67 | ||
XM_023644103.1 | XP_023499871.1 | 433 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
