Assay Details
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637564.1 | XP_023493332.1 | 1973 | 58 | ||
XM_023637565.1 | XP_023493333.1 | 1808 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
