Assay Details
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001914831.5 | XP_001914866.1 | 716 | 65 | ||
XM_023637676.1 | XP_023493444.1 | 667 | 65 | |||
XM_023637677.1 | XP_023493445.1 | 1240 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
