Assay Details
Gene Symbol: HERC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637736.1 | XP_023493504.1 | 1186 | 70 | ||
XM_023637737.1 | XP_023493505.1 | 1187 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
