Assay Details
Gene Symbol: ENOPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493012.6 | XP_001493062.1 | 426 | 66 | ||
XM_023637846.1 | XP_023493614.1 | 310 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
