Assay Details
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637908.1 | XP_023493676.1 | 1097 | 67 | ||
XM_023637909.1 | XP_023493677.1 | 988 | 67 | |||
XM_023637910.1 | XP_023493678.1 | 1056 | 67 | |||
XM_023637911.1 | XP_023493679.1 | 927 | 67 | |||
GenBank mRNA | JL619335.1 | - | 2 - 3 | 177 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
