Assay Details
Gene Symbol: AREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489473.4 | XP_001489523.1 | 2857 | 67 | ||
XM_014738864.2 | XP_014594350.1 | 762 | 67 | |||
XM_023637967.1 | XP_023493735.1 | 2790 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
