Assay Details
Gene Symbol: COX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488484.5 | XP_001488534.3 | 561 | 138 | ||
XM_005608725.3 | XP_005608782.2 | 560 | 138 | |||
XM_014738806.2 | XP_014594292.1 | 247 | 138 | |||
GenBank mRNA | JL622018.1 | - | 2 - 3 | 227 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014738806.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
