Assay Details
Gene Symbol: CENPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497321.6 | XP_001497371.3 | 584 | 65 | ||
XM_023638031.1 | XP_023493799.1 | 460 | 65 | |||
XM_023638032.1 | XP_023493800.1 | 534 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023638032.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
