Assay Details
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608749.3 | XP_005608806.1 | 1423 | 73 | ||
XM_005608750.3 | XP_005608807.1 | 1378 | 73 | |||
XM_005608751.3 | XP_005608808.1 | 1480 | 73 | |||
XM_005608752.3 | XP_005608809.1 | 1147 | 73 | |||
XM_014738728.2 | XP_014594214.1 | 1160 | 73 | |||
XM_014738729.2 | XP_014594215.1 | 524 | 73 | |||
XM_023638056.1 | XP_023493824.1 | 1605 | 73 | |||
XM_023638057.1 | XP_023493825.1 | 1421 | 73 | |||
XM_023638058.1 | XP_023493826.1 | 1329 | 73 | |||
XM_023638059.1 | XP_023493827.1 | 1483 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
