Assay Details
Gene Symbol: SRD5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916943.5 | XP_001916978.4 | 653 | 94 | ||
GenBank mRNA | JL619792.1 | - | 2 - 3 | 332 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
