Assay Details
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493770.4 | XP_001493820.1 | 1150 | 59 | ||
XM_005608775.3 | XP_005608832.1 | 1154 | 59 | |||
XM_023638098.1 | XP_023493866.1 | 1153 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
